Thermo Scientific Chemicals

Ethyl cyanoformate, 99%, Thermo Scientific Chemicals

Catalog number: L15777.22
100 g, Each
Thermo Scientific Chemicals

Ethyl cyanoformate, 99%, Thermo Scientific Chemicals

Catalog number: L15777.22
100 g, Each
Quantity
Catalog number: L15777.22
also known as L15777-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
623-49-4
IUPAC Name
ethyl cyanoformate
Molecular Formula
C4H5NO2
InChI Key
MSMGXWFHBSCQFB-UHFFFAOYSA-N
SMILES
CCOC(=O)C#N
Assay (GC)
≥98.5%
Comment
May form slight precipitate on storage
Identification (FTIR)
Conforms
Refractive Index
1.3800-1.3840 @ 20?C
Appearance (Color)
Clear or slightly turbid colorless

Description

Ethyl cyanoformate is used as reagent in the preparation of N-substituted amindinoformic acid and ethyl-4-quinazoline -2-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyanoformate is used as reagent in the preparation of N-substituted amindinoformic acid and ethyl-4-quinazoline -2-carboxylate.

Solubility
Miscible with chloroform and methanol.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text