Thermo Scientific Chemicals

2-(4-Aminophenyl)ethylamine, 98%, Thermo Scientific Chemicals

Catalog number: L15782.06
5 g, Each
Thermo Scientific Chemicals

2-(4-Aminophenyl)ethylamine, 98%, Thermo Scientific Chemicals

Catalog number: L15782.06
5 g, Each
Quantity
Catalog number: L15782.06
also known as L15782-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13472-00-9
IUPAC Name
4-(2-aminoethyl)aniline
Molecular Formula
C8H12N2
InChI Key
LNPMZQXEPNWCMG-UHFFFAOYSA-N
SMILES
NCCC1=CC=C(N)C=C1
Appearance (Color)
White or colorless to pale yellow or yellow-brown
Form
Low melting fused solid clear as liquid
Assay from Supplier's CofA
≥97.5% (GC)

Description

4-(2-Aminoethyl)aniline has been used in chemical modification of silk fibroin to tailor the overall hydrophilicity and structure of silk. It has been used as reagent in polycondensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Aminoethyl)aniline has been used in chemical modification of silk fibroin to tailor the overall hydrophilicity and structure of silk. It has been used as reagent in polycondensation reactions.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text