Dimethyl sebacate, 97%, Thermo Scientific Chemicals
Dimethyl sebacate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl sebacate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L15805.22
also known as L15805-22
Price (USD)/ Each
53.30
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
53.30
Add to cart
Dimethyl sebacate, 97%, Thermo Scientific Chemicals
Catalog numberL15805.22
Price (USD)/ Each
53.30
-
Add to cart
Chemical Identifiers
CAS106-79-6
IUPAC Name1,10-dimethyl decanedioate
Molecular FormulaC12H22O4
InChI KeyALOUNLDAKADEEB-UHFFFAOYSA-N
SMILESCOC(=O)CCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4360-1.4410 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Polytetramethylene sebacate was prepared from dimethyl sebacate. Dimethyl sebacate was used in the biosynthesis of oligomeric sebacates as lubricant oils. Dimethyl sebacate was a convenient starting material for cyclization of N-Succinylglycine dimethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polytetramethylene sebacate was prepared from dimethyl sebacate. Dimethyl sebacate was used in the biosynthesis of oligomeric sebacates as lubricant oils. Dimethyl sebacate was a convenient starting material for cyclization of N-Succinylglycine dimethyl ester.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. S. Gryglewicz,; F. A. Oko. Synthesis and Biosynthesis of Oligomeric Sebacates as Lubricant Oils. nd. Eng. Chem. Res. 2005, volume (6),1640-1644.
  2. Harold Werbin,; Paul E. Spoerri. Cyclization of N-Succinylglycine Dimethyl Ester. J. Am. Chem. Soc. 1947 , 69 (7), 1681-1684.