6-Methoxy-1-indanone, 99%, Thermo Scientific Chemicals
6-Methoxy-1-indanone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methoxy-1-indanone, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number L15819.06
also known as L15819-06
Price (USD)/ Each
134.65
Online exclusive
149.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
134.65
Online exclusive
149.00 
Save 14.35 (10%)
Add to cart
6-Methoxy-1-indanone, 99%, Thermo Scientific Chemicals
Catalog numberL15819.06
Price (USD)/ Each
134.65
Online exclusive
149.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13623-25-1
IUPAC Name6-methoxy-2,3-dihydro-1H-inden-1-one
Molecular FormulaC10H10O2
InChI KeyUJGDLLGKMWVCPT-UHFFFAOYSA-N
SMILESCOC1=CC=C2CCC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)104.0-110.0?C
FormPowder
Appearance (Color)Cream
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only