Thermo Scientific Chemicals

Zinc cyanide, 98%, Thermo Scientific Chemicals

Catalog number: L15836.0B
1000 g, Each
Thermo Scientific Chemicals

Zinc cyanide, 98%, Thermo Scientific Chemicals

Catalog number: L15836.0B
1000 g, Each
Quantity
Catalog number: L15836.0B
also known as L15836-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-21-1
IUPAC Name
zinc cyanide
Molecular Formula
C2N2Zn
InChI Key
GTLDTDOJJJZVBW-UHFFFAOYSA-N
SMILES
[Zn++].[C-]#N.[C-]#N
Assay (unspecified)
>97.5% by Zn EA
Elemental Analysis
Zn: 54.58-56.80% (Theory 55.69%)
Appearance (Color)
White
Form
Powder

Description

Zinc cyanide is used for electroplating of zinc. In Gatterman reaction , it is used for the introduction of formyl group in to aromatic compounds as well as to prepare 2-hydroxy-1-nafthaldehyde and mesitaldehyde. It is also used in chemical analysis in analytical chemistry. Further, it acts as a catalyst for the cyanosilylation of aldehydes and ketones. In addition to this, it reacts with 1,3,5-triethyl-benzene to get 2,4,6-triethyl-benzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc cyanide is used for electroplating of zinc. In Gatterman reaction , it is used for the introduction of formyl group in to aromatic compounds as well as to prepare 2-hydroxy-1-nafthaldehyde and mesitaldehyde. It is also used in chemical analysis in analytical chemistry. Further, it acts as a catalyst for the cyanosilylation of aldehydes and ketones. In addition to this, it reacts with 1,3,5-triethyl-benzene to get 2,4,6-triethyl-benzaldehyde.

Solubility
Soluble in alkalies, potassium cyanide and ammonia. Insoluble in water and most solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and acids.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text