Ethyl 4-aminopiperidine-1-carboxylate, 98%, Thermo Scientific Chemicals
Ethyl 4-aminopiperidine-1-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-aminopiperidine-1-carboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L15866.06
also known as L15866-06
Price (USD)/ Each
36.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
36.80
Add to cart
Ethyl 4-aminopiperidine-1-carboxylate, 98%, Thermo Scientific Chemicals
Catalog numberL15866.06
Price (USD)/ Each
36.80
-
Add to cart
Chemical Identifiers
CAS58859-46-4
IUPAC Name1-(ethoxycarbonyl)piperidin-4-aminium
Molecular FormulaC8H17N2O2
InChI KeyGQQQULCEHJQUJT-UHFFFAOYSA-O
SMILESCCOC(=O)N1CCC([NH3+])CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4805-1.4845 @ 20?C
FormLiquid
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only