Thermo Scientific Chemicals

5-Methylthiazole, 97%, Thermo Scientific Chemicals

Catalog number: L15879.06
5 g, Each
Thermo Scientific Chemicals

5-Methylthiazole, 97%, Thermo Scientific Chemicals

Catalog number: L15879.06
5 g, Each
Quantity
Catalog number: L15879.06
also known as L15879-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
3581-89-3
IUPAC Name
5-methyl-1,3-thiazole
Molecular Formula
C4H5NS
InChI Key
RLYUNPNLXMSXAX-UHFFFAOYSA-N
SMILES
CC1=CN=CS1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5265-1.5315 @ 20?C

Description

5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to yeild mobile, room-temperature ionic liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-methyl thiazole undergoes alkylation with n-butyl bromide followed by anion exchange of Br- for BF4- to yeild mobile, room-temperature ionic liquids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention