4-Amino-2-chlorobenzonitrile is commonly used as an organic reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-2-chlorobenzonitrile is commonly used as an organic reagent.
Notes
Incompatible with acid chlorides, acid anhydrides and chloroformates.
RUO – Research Use Only
Peukert, S.; Hughes, R.; Nunez, J.; He, G.; Yan, Z.; Jain, R.; Llamas, L.; Luchansky, S.; Carlson, A.; Liang, G.; Kunjathoor, V.; Pietropaolo, M.; Shapiro, J.; Castellana, A.; Wu, X.; Bose, A. Discovery of 2-Pyridylpyrimidines as the First Orally Bioavailable GPR39 Agonists. ACS Med. Chem. Lett. 2014, 5 (10), 1114-1118.
Kumar, V.; Rachamalla, M.; Nandekar, P.; Khatik, G. L.; Sangamwar, A. T.; Tikoo, K.; Nair, V. A. Design and synthesis of optically pure 3-aryl-6-methyl-2-thioxotetrahydropyrimidin-4(1H)-ones as anti-prostate cancer agents. RSC Adv. 2014, 4 (71), 37868-37877.