Thermo Scientific Chemicals

4-Amino-2-chlorobenzonitrile, 97+%, Thermo Scientific Chemicals

Catalog number: L15924.14
25 g, Each
Thermo Scientific Chemicals

4-Amino-2-chlorobenzonitrile, 97+%, Thermo Scientific Chemicals

Catalog number: L15924.14
25 g, Each
Quantity
Catalog number: L15924.14
also known as L15924-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20925-27-3
IUPAC Name
4-amino-2-chlorobenzonitrile
Molecular Formula
C7H5ClN2
InChI Key
ZFBKYGFPUCUYIF-UHFFFAOYSA-N
SMILES
NC1=CC=C(C#N)C(Cl)=C1
Form
Powder or lumps
Appearance (Color)
Yellow to pale brown
Assay (GC)
≥97.0%
Melting Point (clear melt)
114.0-121.0?C

Description

4-Amino-2-chlorobenzonitrile is commonly used as an organic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2-chlorobenzonitrile is commonly used as an organic reagent.

Notes
Incompatible with acid chlorides, acid anhydrides and chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text