D-(-)-Pantolactone, 99%, Thermo Scientific Chemicals
D-(-)-Pantolactone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-(-)-Pantolactone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L15983.14
also known as L15983-14
Price (USD)/ Each
64.50
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
64.50
Add to cart
D-(-)-Pantolactone, 99%, Thermo Scientific Chemicals
Catalog numberL15983.14
Price (USD)/ Each
64.50
-
Add to cart
Chemical Identifiers
CAS599-04-2
IUPAC Name3-hydroxy-4,4-dimethyloxolan-2-one
Molecular FormulaC6H10O3
InChI KeySERHXTVXHNVDKA-UHFFFAOYNA-N
SMILESCC1(C)COC(=O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)88-93?C
Optical Rotation-48? to -52? (c=2 in water)
Appearance (Color)White
FormCrystalline powder or crystals
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only