Thermo Scientific Chemicals

2,6-Dimethyl-4-heptanone, >90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone), Thermo Scientific Chemicals

Catalog number: L16015.AP
500 mL, Each
Thermo Scientific Chemicals

2,6-Dimethyl-4-heptanone, >90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone), Thermo Scientific Chemicals

Catalog number: L16015.AP
500 mL, Each
Quantity
Catalog number: L16015.AP
also known as L16015-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-83-8
IUPAC Name
2,6-dimethylheptan-4-one
Molecular Formula
C9H18O
InChI Key
PTTPXKJBFFKCEK-UHFFFAOYSA-N
SMILES
CC(C)CC(=O)CC(C)C
Assay (GC)
≥90.0% (sum of 2,6-Dimethyl-4-heptanone & 4,6-dimethyl-2-heptanone, non-U.S. specification)
Assay from Suppliers CofA
≥90.0% (U.S. specification)
Refractive Index
1.4100-1.4180 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

2,6-Dimethyl-4-heptanone is used as a coating solvent. It is an active component of mint oil. It acts as a dispersant for organosol type resins. It is involved in the antigerminative treatment of bulbs and tubers. Further, it is used as a solvent for nitrocellulose. In addition to this, it acts as an intermediate in the preparation of inhibitors, active pharmaceutical ingredients and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-4-heptanone is used as a coating solvent. It is an active component of mint oil. It acts as a dispersant for organosol type resins. It is involved in the antigerminative treatment of bulbs and tubers. Further, it is used as a solvent for nitrocellulose. In addition to this, it acts as an intermediate in the preparation of inhibitors, active pharmaceutical ingredients and dyes.

Solubility
Miscible with ethanol, ether, carbon tetrachloride, chloroform, benzene and most organic liquids.

Notes
Incompatible with strong bases, strong oxidizing agents, strong acids and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text