Thermo Scientific Chemicals

Methyl indole-6-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: L16020.06
5 g, Each
Thermo Scientific Chemicals

Methyl indole-6-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: L16020.06
5 g, Each
Quantity
Catalog number: L16020.06
also known as L16020-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50820-65-0
IUPAC Name
methyl 1H-indole-6-carboxylate
Molecular Formula
C10H9NO2
InChI Key
AYYOZKHMSABVRP-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C2C=CNC2=C1
Appearance (Color)
White to pale cream to cream to yellow to orange to brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text