Thermo Scientific Chemicals

Lithium amide, 95%, Thermo Scientific Chemicals

Catalog number: L16023.14
25 g, Each
Thermo Scientific Chemicals

Lithium amide, 95%, Thermo Scientific Chemicals

Catalog number: L16023.14
25 g, Each
Quantity
Catalog number: L16023.14
also known as L16023-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7782-89-0
IUPAC Name
lithium(1+) azanide
Molecular Formula
H2LiN
InChI Key
AFRJJFRNGGLMDW-UHFFFAOYSA-N
SMILES
[Li+].[NH2-]
Appearance (Color)
White to grey
Form
Powder (may contain black specks)
Assay (unspecified)
≥94.0%

Description

Lithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.

Solubility
Slightly soluble in ethanol and liquid ammonia. Insoluble in anhydrous ether, benzene and toluene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, water and alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text