gamma-Undecanolactone, 99%, Thermo Scientific Chemicals
gamma-Undecanolactone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

gamma-Undecanolactone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number L16028.AP
also known as L16028-AP
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
gamma-Undecanolactone, 99%, Thermo Scientific Chemicals
Catalog numberL16028.AP
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS104-67-6
IUPAC Name(5S)-5-heptyloxolan-2-one
Molecular FormulaC11H20O2
InChI KeyPHXATPHONSXBIL-JTQLQIEISA-N
SMILESCCCCCCC[C@H]1CCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4490-1.4520 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only