4-(Trifluoromethylthio)aniline, 98%, Thermo Scientific Chemicals
4-(Trifluoromethylthio)aniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethylthio)aniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L16067.06
also known as L16067-06
Price (USD)/ Each
166.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
166.00
Add to cart
4-(Trifluoromethylthio)aniline, 98%, Thermo Scientific Chemicals
Catalog numberL16067.06
Price (USD)/ Each
166.00
-
Add to cart
Chemical Identifiers
CAS372-16-7
IUPAC Name4-[(trifluoromethyl)sulfanyl]aniline
Molecular FormulaC7H6F3NS
InChI KeyOHHHTUXVBNGOGI-UHFFFAOYSA-N
SMILESNC1=CC=C(SC(F)(F)F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Refractive Index1.5255-1.5295 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only