Thermo Scientific Chemicals

(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L16086.06
5 g, Each
Thermo Scientific Chemicals

(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L16086.06
5 g, Each
Quantity
Catalog number: L16086.06
also known as L16086-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78266-06-5
IUPAC Name
2-({[(3-bromo-2,4,6-trimethylphenyl)carbamoyl]methyl}(carboxymethyl)amino)acetic acid
Molecular Formula
C15H19BrN2O5
InChI Key
MHPZZZZLAQGTHT-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(NC(=O)CN(CC(O)=O)CC(O)=O)C(C)=C1Br
Form
Powder or lumps or fused solid
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Appearance (Color)
White
Assay (HPLC)
≥98.0%

Description

(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.

Solubility
Soluble in dimethylsulphoxide and ethanol. Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text