Thermo Scientific Chemicals

Bis(pinacolato)diboron, 98+%, Thermo Scientific Chemicals

Catalog number: L16088.06
5 g, Each
Thermo Scientific Chemicals

Bis(pinacolato)diboron, 98+%, Thermo Scientific Chemicals

Catalog number: L16088.06
5 g, Each
Quantity
Catalog number: L16088.06
also known as L16088-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73183-34-3
IUPAC Name
4,4,5,5-tetramethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
Molecular Formula
C12H24B2O4
InChI Key
IPWKHHSGDUIRAH-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)B1OC(C)(C)C(C)(C)O1
Appearance (Color)
White to pale cream
Form
Lumps and/or powder
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms (UK sourced material only)
Proton NMR
Conforms to structure (US sourced material only)

Description

Bis(pinacolato)diboron is a reagent used for the cis-vicinal diborylation of acetylenes and olefins with platinum catalysis. It acts as a substrate used in a new palladium-catalyzed cyclization of 1,6-enynes leading to homoallylic alkylboronates. It is used as a condensation agent in the preparation of poly(arylene)s. It plays an essential role in Suzuki reaction and used as matrix metallo-proteinase (MMP-2) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(pinacolato)diboron is a reagent used for the cis-vicinal diborylation of acetylenes and olefins with platinum catalysis. It acts as a substrate used in a new palladium-catalyzed cyclization of 1,6-enynes leading to homoallylic alkylboronates. It is used as a condensation agent in the preparation of poly(arylene)s. It plays an essential role in Suzuki reaction and used as matrix metallo-proteinase (MMP-2) inhibitor.

Solubility
Soluble in tetrahydrofuran, dichloromethane, toluene, hexane and heptane. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text