Thermo Scientific Chemicals

6-Bromo-2-naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L16093.14
25 g, Each
Thermo Scientific Chemicals

6-Bromo-2-naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: L16093.14
25 g, Each
Quantity
Catalog number: L16093.14
also known as L16093-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5773-80-8
IUPAC Name
6-bromonaphthalene-2-carboxylic acid
Molecular Formula
C11H7BrO2
InChI Key
NPMCAVBMOTZUPD-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C2C=C(Br)C=CC2=C1
Appearance (Color)
White to cream to brown
Form
Powder or lumps
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (HPLC)
≥97.5%

Description

Benzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzimidazole derivatives has been prepared by using ortho-phenyl diamine and 6-bromo-2-naphthoic acid.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text