Thermo Scientific Chemicals

Amberlite XAD-1180, Thermo Scientific Chemicals

Catalog number: L16097.36
500 g, Each
Thermo Scientific Chemicals

Amberlite XAD-1180, Thermo Scientific Chemicals

Catalog number: L16097.36
500 g, Each
Quantity
Catalog number: L16097.36
also known as L16097-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97396-56-0
Appearance (Color)
White
Form
beads
Comment
Surface area: >550 m2/g
Comment
Uniformity coefficient: 0.00 - 1.80
Harmonic mean size
0.350 - 0.600 mm

Description

Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymeric adsorbent for hydrophobic compounds: antibiotics, vitamins, amino acids, enzymes; for purification: gallium, myxovirescins, phosphoric acid, riboflavin salts. Amberlite™ XAD1180N can be used as a polymeric adsorbent. The regeneration of this adsorbent has been effective in alkaline medium and tests have shown the adsorbent efficiently removes chromium from tannery waste water. Mainly used in the Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound, Amberlite XAD-1180 is a polymeric adsorbent resin that is used primarily in the purification and preparation of hydrophobic compound.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text