Thermo Scientific Chemicals

Isopropyl isocyanate, 98+%, Thermo Scientific Chemicals

Catalog number: L16105.03
1 g, Each
Thermo Scientific Chemicals

Isopropyl isocyanate, 98+%, Thermo Scientific Chemicals

Catalog number: L16105.03
1 g, Each
Quantity
Catalog number: L16105.03
also known as L16105-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1795-48-8
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.3820-1.3860 @ 20?C

Description

Isopropyl isocyanate is used as a reagent in the preparation of androgen receptor antagonist for prostate cancer treatment. It is also used in the synthesis of selective dual inhibitor of mammalian target of rapamycin (mTORC1 and mTORC2). Further, it serves as a reagent in the synthesis of O-aryl N-isopropylcarbamates. In addition to this, it is used as a derivatization reagent during the stereoisomeric analysis of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl isocyanate is used as a reagent in the preparation of androgen receptor antagonist for prostate cancer treatment. It is also used in the synthesis of selective dual inhibitor of mammalian target of rapamycin (mTORC1 and mT
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text