Thermo Scientific Chemicals

4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: L16150.14
25 g, Each
Thermo Scientific Chemicals

4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%, Thermo Scientific Chemicals

Catalog number: L16150.14
25 g, Each
Quantity
Catalog number: L16150.14
also known as L16150-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1107-00-2
IUPAC Name
5-[2-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-1,3-dihydro-2-benzofuran-1,3-dione
Molecular Formula
C19H6F6O6
InChI Key
QHHKLPCQTTWFSS-UHFFFAOYSA-N
SMILES
FC(F)(F)C(C1=CC=C2C(=O)OC(=O)C2=C1)(C1=CC=C2C(=O)OC(=O)C2=C1)C(F)(F)F
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
241-248?C
Appearance (Color)
White to cream
Form
Powder

Description

4,4'-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text