1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals
1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L16185.14
also known as L16185-14
Price (USD)/ Each
81.65
Online exclusive
90.70 
Save 9.05 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
81.65
Online exclusive
90.70 
Save 9.05 (10%)
Add to cart
1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals
Catalog numberL16185.14
Price (USD)/ Each
81.65
Online exclusive
90.70 
Save 9.05 (10%)
-
Add to cart
Chemical Identifiers
CAS54-95-5
IUPAC Name5H,6H,7H,8H,9H-[1,2,3,4]tetrazolo[1,5-a]azepine
Molecular FormulaC6H10N4
InChI KeyCWRVKFFCRWGWCS-UHFFFAOYSA-N
SMILESC1CCN2N=NN=C2CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)56-62?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only