Thermo Scientific Chemicals

Chlorodimethylvinylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L16198.22
100 g, Each
Thermo Scientific Chemicals

Chlorodimethylvinylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L16198.22
100 g, Each
Quantity
Catalog number: L16198.22
also known as L16198-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1719-58-0
IUPAC Name
[(1E)-2-chloroethenyl]dimethylsilyl
Molecular Formula
C4H8ClSi
InChI Key
RABBDIBWJVOAPB-ONEGZZNKSA-N
SMILES
C[Si](C)C=CCl
Appearance (Color)
Clear colorless to white to yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4135-1.4185 @ 20?C
Identification (FTIR)
Conforms

Description

Chlorodimethylvinylsilane is used to prepare silicon-containing polymers, silaheterocycles and new chelating ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylvinylsilane is used to prepare silicon-containing polymers, silaheterocycles and new chelating ligands.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text