5-Amino-2-nitrobenzoic acid, 95%, Thermo Scientific Chemicals
5-Amino-2-nitrobenzoic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Amino-2-nitrobenzoic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16225.06
also known as L16225-06
Price (USD)/ Each
83.65
Online exclusive
92.30 
Save 8.65 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
83.65
Online exclusive
92.30 
Save 8.65 (9%)
Add to cart
5-Amino-2-nitrobenzoic acid, 95%, Thermo Scientific Chemicals
Catalog numberL16225.06
Price (USD)/ Each
83.65
Online exclusive
92.30 
Save 8.65 (9%)
-
Add to cart
Chemical Identifiers
CAS13280-60-9
IUPAC Name5-amino-2-nitrobenzoic acid
Molecular FormulaC7H6N2O4
InChI KeyKZZWQCKYLNIOBT-UHFFFAOYSA-N
SMILESNC1=CC=C(C(=C1)C(O)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥94.0%
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only