Thermo Scientific Chemicals

Methyl indole-5-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: L16228.03
1 g, Each
Thermo Scientific Chemicals

Methyl indole-5-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: L16228.03
1 g, Each
Quantity
Catalog number: L16228.03
also known as L16228-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1011-65-0
IUPAC Name
methyl 1H-indole-5-carboxylate
Molecular Formula
C10H9NO2
InChI Key
DRYBMFJLYYEOBZ-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C2NC=CC2=C1
Form
Powder
Appearance (Color)
White to pale cream to pale yellow
Assay (GC)
≥96.0%
Melting Point (clear melt)
119-130°C

Description

Reactant for biosynthesis of inhibitors of protein kinases, reactant in metal-free Friedel-Crafts alkylation, reactant in preparation of diphenylsulfonium ylides from Martin?s sulfurane, reactant in cross dehydrogenative coupling reactions, reactant in synthesis of indirubin derivatives, reactant in preparation of aminoindolylacetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for biosynthesis of inhibitors of protein kinases, reactant in metal-free Friedel-Crafts alkylation, reactant in preparation of diphenylsulfonium ylides from Martin′s sulfurane, reactant in cross dehydrogenative coupling reactions, reactant in synthesis of indirubin derivatives, reactant in preparation of aminoindolylacetates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text