Thermo Scientific Chemicals

3-(2-Aminoethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: L16311.03
1 g, Each
Thermo Scientific Chemicals

3-(2-Aminoethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: L16311.03
1 g, Each
Quantity
Catalog number: L16311.03
also known as L16311-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20173-24-4
IUPAC Name
2-(pyridin-3-yl)ethan-1-aminium
Molecular Formula
C7H11N2
InChI Key
NAHHNSMHYCLMON-UHFFFAOYSA-O
SMILES
[NH3+]CCC1=CC=CN=C1
Comment
Material sourced in the U.S. and in other countries
Refractive Index
1.5380-1.5420 @ 20°C (non-U.S. sourced material)
Infrared spectrum
Conforms (U.S. sourced material)
Appearance (Color)
Clear colorless to pale yellow to brown
Assay (GC)
≥97.5%

Description

3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text