3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
RUO – Research Use Only
General References:
- Małgorzata Dukat,; Anna Ramunno,; Rita Banzi,; M. Imad Damaj,; Billy Martin,; Richard A. Glennon. 3-(2-Aminoethyl)pyridine analogs as α4β2 nicotinic cholinergic receptor ligands. Journal name. 2005, 15(19), 4308-4312.