(R)-(+)-1-Phenylpropylamine, ChiPros 99+%, ee 98%, Thermo Scientific Chemicals
(R)-(+)-1-Phenylpropylamine, ChiPros 99+%, ee 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(+)-1-Phenylpropylamine, ChiPros 99+%, ee 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16319.06
also known as L16319-06
Price (USD)/ Each
65.65
Online exclusive
73.10 
Save 7.45 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
65.65
Online exclusive
73.10 
Save 7.45 (10%)
Add to cart
(R)-(+)-1-Phenylpropylamine, ChiPros 99+%, ee 98%, Thermo Scientific Chemicals
Catalog numberL16319.06
Price (USD)/ Each
65.65
Online exclusive
73.10 
Save 7.45 (10%)
-
Add to cart
Chemical Identifiers
CAS3082-64-2
IUPAC Name1-phenylpropan-1-amine
Molecular FormulaC9H13N
InChI KeyAQFLVLHRZFLDDV-UHFFFAOYNA-N
SMILESCCC(N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥99.0%
Enantiomeric excess≥97.5%
Optical Rotation20? ± 0.5? (neat)
View more
For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only