Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.
Solubility
Soluble in water (10g/L).
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only
Hu, Meng; Zhang, Fu-Li; Xie, Mei-Hua. A Simple and Efficient Synthesis of (S)- and (R)-1-(3-Methoxyphenyl) Ethylamine. Letters in Organic Chemistry. 2007, 4(2), 126-128(3).