Thermo Scientific Chemicals

3-Pyrroline, 96%, Thermo Scientific Chemicals

Catalog number: L16349.03
1 g, Each
Thermo Scientific Chemicals

3-Pyrroline, 96%, Thermo Scientific Chemicals

Catalog number: L16349.03
1 g, Each
Quantity
Catalog number: L16349.03
also known as L16349-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-96-6
IUPAC Name
2,5-dihydro-1H-pyrrole
Molecular Formula
C4H7N
InChI Key
JVQIKJMSUIMUDI-UHFFFAOYSA-N
SMILES
C1NCC=C1
Appearance (Color)
Clear colorless
Refractive Index
1.4630-1.4715 @ 20?C
Form
Liquid
Assay (GC)
≥92.0%

Description

3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.

Solubility
Fully misicble in water.

Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text