3-Bromobenzeneboronic acid, 98+%, Thermo Scientific Chemicals
3-Bromobenzeneboronic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromobenzeneboronic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16354.06
also known as L16354-06
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
3-Bromobenzeneboronic acid, 98+%, Thermo Scientific Chemicals
Catalog numberL16354.06
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS89598-96-9
IUPAC Name(3-bromophenyl)boronic acid
Molecular FormulaC6H6BBrO2
InChI KeyAFSSVCNPDKKSRR-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)≥98.0%
Assay (HPLC)≥98.0%
Proton NMRConforms to structure
Appearance (Color)White to cream to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only