2,4-Dichloro-6-methylbenzonitrile, 97%, Thermo Scientific Chemicals
2,4-Dichloro-6-methylbenzonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dichloro-6-methylbenzonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number L16372.06
also known as L16372-06
Price (USD)/ Each
312.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
312.00
Add to cart
2,4-Dichloro-6-methylbenzonitrile, 97%, Thermo Scientific Chemicals
Catalog numberL16372.06
Price (USD)/ Each
312.00
-
Add to cart
Chemical Identifiers
CAS175277-98-2
IUPAC Name2,4-dichloro-6-methylbenzonitrile
Molecular FormulaC8H5Cl2N
InChI KeyNFPYAMXNKNDVDS-UHFFFAOYSA-N
SMILESCC1=CC(Cl)=CC(Cl)=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to brown
FormCrystals or powder or crystalline powder or fused solid or granules
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)72.0-82.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only