Thermo Scientific Chemicals

Ethyl pyrrole-2-carboxylate, 98+%, Thermo Scientific Chemicals

Catalog number: L16382.06
5 g, Each
Thermo Scientific Chemicals

Ethyl pyrrole-2-carboxylate, 98+%, Thermo Scientific Chemicals

Catalog number: L16382.06
5 g, Each
Quantity
Catalog number: L16382.06
also known as L16382-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2199-43-1
IUPAC Name
ethyl 1H-pyrrole-2-carboxylate
Molecular Formula
C7H9NO2
InChI Key
PAEYAKGINDQUCT-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CN1
Appearance (Color)
Colorless to white to cream to orange to brown
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder or lumps or fused solid or clear liquid as melt

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text