Thermo Scientific Chemicals

3-(Acryloyloxy)propyltrimethoxysilane, 94%, stab. with 100ppm BHT, Thermo Scientific Chemicals

Catalog number: L16400.14
25 g, Each
Thermo Scientific Chemicals

3-(Acryloyloxy)propyltrimethoxysilane, 94%, stab. with 100ppm BHT, Thermo Scientific Chemicals

Catalog number: L16400.14
25 g, Each
Quantity
Catalog number: L16400.14
also known as L16400-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4369-14-6
IUPAC Name
3-(trimethoxysilyl)propyl prop-2-enoate
Molecular Formula
C9H18O5Si
InChI Key
KBQVDAIIQCXKPI-UHFFFAOYSA-N
SMILES
CO[Si](CCCOC(=O)C=C)(OC)OC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥92.5%
Identification (FTIR)
Conforms
Refractive Index
1.4255-1.4325 @ 20?C

Description

3-(Acryloyloxy)propyltrimethoxysilane is used to enhance the thermal conductivity of silicone rubber filled with hybrid fillers. It is involved in the synthesis of vinyl and acryalte modified silica nanoparticles by using vinyltrimethoxysilane. Further, it is used as a silane coupling agent involved in the modification of fluorescein isothiocyanate-mesoporous silica nanoparticles with carbon-carbon double bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Acryloyloxy)propyltrimethoxysilane is used to enhance the thermal conductivity of silicone rubber filled with hybrid fillers. It is involved in the synthesis of vinyl and acryalte modified silica nanoparticles by using vinyltrimethoxysilane. Further, it is used as a silane coupling agent involved in the modification of fluorescein isothiocyanate-mesoporous silica nanoparticles with carbon-carbon double bonds.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids and bases. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text