(3-Aminopropyl)diethoxymethylsilane, 97%, Thermo Scientific Chemicals
(3-Aminopropyl)diethoxymethylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(3-Aminopropyl)diethoxymethylsilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L16401.14
also known as L16401-14
Price (USD)/ Each
29.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
29.30
Add to cart
(3-Aminopropyl)diethoxymethylsilane, 97%, Thermo Scientific Chemicals
Catalog numberL16401.14
Price (USD)/ Each
29.30
-
Add to cart
Chemical Identifiers
CAS3179-76-8
IUPAC Name(3-aminopropyl)diethoxymethylsilane
Molecular FormulaC8H21NO2Si
InChI KeyHXLAEGYMDGUSBD-UHFFFAOYSA-N
SMILESCCO[Si](C)(CCCN)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
FormLiquid
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

Solubility
Soluble and reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Young-Hye La,; Hyun Ju Kim,; Il Sang Maeng,; Yu Jin Jung,;Joon Won Park. Selective Cleavage of Nitro Groups in Nitro-Substituted Aromatic Monolayers by Synchrotron Soft X-rays:  Effect of Molecular Structure on Cleavage Rates. Langmuir. 2002, 18 (6), 2430-2433.