Thermo Scientific Chemicals

(3-Aminopropyl)diethoxymethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L16401.22
100 g, Each
Thermo Scientific Chemicals

(3-Aminopropyl)diethoxymethylsilane, 97%, Thermo Scientific Chemicals

Catalog number: L16401.22
100 g, Each
Quantity
Catalog number: L16401.22
also known as L16401-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3179-76-8
IUPAC Name
(3-aminopropyl)diethoxymethylsilane
Molecular Formula
C8H21NO2Si
InChI Key
HXLAEGYMDGUSBD-UHFFFAOYSA-N
SMILES
CCO[Si](C)(CCCN)OCC
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Form
Liquid

Description

(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

Solubility
Soluble and reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text