Thermo Scientific Chemicals

4-(Chloromethyl)phenyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: L16431.06
5 g, Each
Thermo Scientific Chemicals

4-(Chloromethyl)phenyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: L16431.06
5 g, Each
Quantity
Catalog number: L16431.06
also known as L16431-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13688-90-9
IUPAC Name
trichloro[4-(chloromethyl)phenyl]silane
Molecular Formula
C7H6Cl4Si
InChI Key
YBYBQFPZMKPGPJ-UHFFFAOYSA-N
SMILES
ClCC1=CC=C(C=C1)[Si](Cl)(Cl)Cl
Appearance (Color)
Clear colorless
Assay from Suppliers CofA
≥96.0%
Appearance (Color)
Clear colorless
Assay from Suppliers CofA
≥96.0%
Form
Liquid

Description

4-(Chloromethyl)phenyltrichlorosilane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Chloromethyl)phenyltrichlorosilane is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents and bases. It is sensitive to moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text