Thermo Scientific Chemicals

Dimethylethoxysilane, 94%, Thermo Scientific Chemicals

Catalog number: L16476.22
100 g, Each
Thermo Scientific Chemicals

Dimethylethoxysilane, 94%, Thermo Scientific Chemicals

Catalog number: L16476.22
100 g, Each
Quantity
Catalog number: L16476.22
also known as L16476-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14857-34-2
IUPAC Name
ethoxydimethylsilyl
Molecular Formula
C4H11OSi
InChI Key
DRUOQOFQRYFQGB-UHFFFAOYSA-N
SMILES
CCO[Si](C)C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Refractive Index
1.3645-1.3715 @ 20?C
Assay (GC)
≥92.0%

Description

Dimethylethoxysilane (DMES), a volatile liquid, is used by NASA to waterproof the heatprotective silica tiles and blankets on the space shuttle. DMES readily produces a variety of products in the presence of small amounts of moisture or bases. DMES readily produces a variety of products in the presence of small amounts of moisture or bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylethoxysilane (DMES), a volatile liquid, is used by NASA to waterproof the heatprotective silica tiles and blankets on the space shuttle. DMES readily produces a variety of products in the presence of small amounts of moisture or bases. DMES readily produces a variety of products in the presence of small amounts of moisture or bases.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text