Thermo Scientific Chemicals

Ethyl dibromofluoroacetate, 97%, Thermo Scientific Chemicals

Catalog number: L16491.06
5 g, Each
Thermo Scientific Chemicals

Ethyl dibromofluoroacetate, 97%, Thermo Scientific Chemicals

Catalog number: L16491.06
5 g, Each
Quantity
Catalog number: L16491.06
also known as L16491-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
565-53-7
IUPAC Name
ethyl 2,2-dibromo-2-fluoroacetate
Molecular Formula
C4H5Br2FO2
InChI Key
COYRDMUQLSIAEX-UHFFFAOYSA-N
SMILES
CCOC(=O)C(F)(Br)Br
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4615-1.4665 @ 20?C

Description

Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dibromofluoroacetate is a reagent used in the preparation of fluorinated molecules in organic synthesis. It is involved in the Reformatsky-type reaction to prepare syn-alfa-bromo-alfa-fluoro-beta-lactams by using diethylzinc. It is also used to prepare 2-fluoroaziridine-2-carboxylates by reaction with imines.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text