Ethyltriethoxysilane, 96%, Thermo Scientific Chemicals
Ethyltriethoxysilane, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyltriethoxysilane, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number L16495.AE
also known as L16495-AE
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
Add to cart
Ethyltriethoxysilane, 96%, Thermo Scientific Chemicals
Catalog numberL16495.AE
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS78-07-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Assay (GC)≥95.0%
Refractive Index1.3890-1.3950 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only