Heptafluoro-2-iodopropane, 97%, stab. with copper powder, Thermo Scientific Chemicals
Heptafluoro-2-iodopropane, 97%, stab. with copper powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Heptafluoro-2-iodopropane, 97%, stab. with copper powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number L16523.22
also known as L16523-22
Price (USD)/ Each
306.65
Online exclusive
341.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
306.65
Online exclusive
341.00 
Save 34.35 (10%)
Add to cart
Heptafluoro-2-iodopropane, 97%, stab. with copper powder, Thermo Scientific Chemicals
Catalog numberL16523.22
Price (USD)/ Each
306.65
Online exclusive
341.00 
Save 34.35 (10%)
-
Add to cart
Chemical Identifiers
CAS677-69-0
IUPAC Name1,1,1,2,3,3,3-heptafluoro-2-iodopropane
Molecular FormulaC3F7I
InChI KeyBBZVTTKMXRPMHZ-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(I)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.3270-1.3390 @ 20°C
Appearance (Color)Clear colorless to pink to purple
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only