(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Junmin Huang.; Hui Chen.; Tingyu Li. Improvement of proline chiral stationary phases by varying peptide length and linker. Journal of Chromatography A. 2006, 1113, (1-2), 109-115.
Franca Bigi.; Andrea Corradini.; Carla Quarantelli.; Giovanni Sartori. Silica-bound decatungstates as heterogeneous catalysts for H2O2 activation in selective sulfide oxidation. Journal of Catalysis. 2007, 250, (2), 222-230.