Thermo Scientific Chemicals

(3-Methylaminopropyl)trimethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: L16541.22
100 g, Each
Thermo Scientific Chemicals

(3-Methylaminopropyl)trimethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: L16541.22
100 g, Each
Quantity
Catalog number: L16541.22
also known as L16541-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3069-25-8
IUPAC Name
methyl[3-(trimethoxysilyl)propyl]amine
Molecular Formula
C7H19NO3Si
InChI Key
DVYVMJLSUSGYMH-UHFFFAOYSA-N
SMILES
CNCCC[Si](OC)(OC)OC
Appearance (Color)
Clear colorless to yellow
Comment
Specification differs for U.S. and non-U.S. material where indicated
Form
Liquid
Refractive Index
1.4170-1.4230 @ 20°C (non-U.S. specification)
Form
Liquid

Description

(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text