2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
Jianfu Ding.; Jia Jiang.; Chantal Blanchetière.; Claire L. Callender. Highly Fluorinated Aromatic-Aliphatic Copolyethers. Macromolecules. 2008, 41 (3), 758-763.
Jianfu Ding.; Yinghua Qi. The Preparation and Characterization of Highly Fluorinated Poly(arylene alkylene ether)s. Macromolecules. 2008, 41 (3), 751-757.