Thermo Scientific Chemicals

1H,1H,2H,2H-Perfluorooctyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: L16606.09
10 g, Each
Thermo Scientific Chemicals

1H,1H,2H,2H-Perfluorooctyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: L16606.09
10 g, Each
Quantity
Catalog number: L16606.09
also known as L16606-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78560-45-9
IUPAC Name
trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Molecular Formula
C8H4Cl3F13Si
InChI Key
PISDRBMXQBSCIP-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.3500-1.3550 @ 20?C

Description

1H,1H,2H,2H-Perfluorooctyltrichlorosilane is used in the silanization of silicon wafer. Self assembled monolayers treated with this, acts as an anti adhesive layer and release of cured poly(dimethyl siloxanes). It is also used to fabricate chemical surface patterns of self assembled monolayers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H,1H,2H,2H-Perfluorooctyltrichlorosilane is used in the silanization of silicon wafer. Self assembled monolayers treated with this, acts as an anti adhesive layer and release of cured poly(dimethyl siloxanes). It is also used to fabricate chemical surface patterns of self assembled monolayers.

Solubility
Miscible with tetrahydrofuran, tetrhydropyran, toluene and other organic solvents.

Notes
Incompatible with water, strong oxidizing agents, alcohols, acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text