Chemical Identifiers
CAS27639-98-1
IUPAC Name2,3,3,3-tetrafluoro-2-{1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy}propanoyl fluoride
Molecular FormulaC12F24O4
InChI KeyIIYQDKPSLAINNX-UHFFFAOYNA-N
SMILESFC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F
View more