Thermo Scientific Chemicals

2,4,6,8-Tetramethylcyclotetrasiloxane, 99%, Thermo Scientific Chemicals

Catalog number: L16642.14
25 g, Each
Thermo Scientific Chemicals

2,4,6,8-Tetramethylcyclotetrasiloxane, 99%, Thermo Scientific Chemicals

Catalog number: L16642.14
25 g, Each
Quantity
Catalog number: L16642.14
also known as L16642-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2370-88-9
Molecular Formula
C4H12O4Si4
InChI Key
WZJUBBHODHNQPW-UHFFFAOYSA-N
SMILES
C[Si]1O[Si](C)O[Si](C)O[Si](C)O1
Molecular Weight (g/mol)
236.48
Assay (GC)
≥98.5%
Refractive Index
1.3855-1.3885 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms

Description

Tetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-cyclotetrasiloxane is a good impregnant of photoelectric material. It is used to manufacture of modified siloxane with defined hydrogen content and chain quantity. It is also used in production of silicone polymers and acts as a precursor for the deposition of polysiloxane, cyclic siloxane, silicon dioxide, oxycarbide thin films with low dielectric constant for microelectronics and semiconductors.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents, bases and acids. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text