2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane is used as an intermediate in organic synthesis. It is used in the preparation of 4-vinyl-benzoic acid ethyl ester. Further, it is used to prepare styrene derivatives by cross-coupling with aryl bromide in the presence of palladium catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane is used as an intermediate in organic synthesis. It is used in the preparation of 4-vinyl-benzoic acid ethyl ester. Further, it is used to prepare styrene derivatives by cross-coupling with aryl bromide in the presence of palladium catalyst.
Solubility
Miscible with most organic solvents.
Notes
Moisture sensitive. Incompatible with acids, bases and strong oxidizing agents.
RUO – Research Use Only
Starting material for carbosilane based spherical dendritic macromolecules: Synthesis, 105 (1998).
Risangud, N.; Li, Z.; Anastasaki, A.; Wilson, P.; Kempe, K.; Haddleton, D. M. Hydrosilylation as an efficient tool for polymer synthesis and modification with methacrylates. RSC Adv. 2015, 5 (8), 5879-5885.
Chen, Q.; de Leon, A.; Advincula, R. C. Inorganic-Organic Thiol-ene Coated Mesh for Oil/Water Separation. ACS Appl. Mater. Interfaces 2015, 7 (33), 18566-18573.