(3,3,3-Trifluoropropyl)methyldichlorosilane, 97%, Thermo Scientific Chemicals
(3,3,3-Trifluoropropyl)methyldichlorosilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(3,3,3-Trifluoropropyl)methyldichlorosilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number L16669.22
also known as L16669-22
Price (USD)/ Each
249.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
249.00
Add to cart
(3,3,3-Trifluoropropyl)methyldichlorosilane, 97%, Thermo Scientific Chemicals
Catalog numberL16669.22
Price (USD)/ Each
249.00
-
Add to cart
Chemical Identifiers
CAS675-62-7
IUPAC Namedichloro(methyl)(3,3,3-trifluoropropyl)silane
Molecular FormulaC4H7Cl2F3Si
InChI KeyOHABWQNEJUUFAV-UHFFFAOYSA-N
SMILESC[Si](Cl)(Cl)CCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.3805-1.3855 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only