(3,3,3-Trifluoropropyl)trichlorosilane, 97%, Thermo Scientific Chemicals
(3,3,3-Trifluoropropyl)trichlorosilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(3,3,3-Trifluoropropyl)trichlorosilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16670.06
also known as L16670-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS592-09-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.3830-1.3880 @ 20?C
(3,3,3-Trifluoropropyl)trichlorosilane is used for the production of fluoro silicones liquids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3,3,3-Trifluoropropyl)trichlorosilane is used for the production of fluoro silicones liquids.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Omar W. Steward.; Ogden R. Pierce. The chlorination of (3,3,3-trifluoropropyl)trichlorosilane. Journal of Organometallic Chemistry. 1965, 4, (2), 138-144.
  2. Paul Tarrant.; G. W. Dyckes.; Robert Dunmire.; G. B. Butler. The Preparation of Some Fluoroalkylmethyldichlorosilanes and their Hydrolysis Products. J. Am. Chem. Soc. 1957, 79, (24), 6536-6540.